4-Chloro-2,3-dihydroinden-1-one 97%

Supplier: Ambeed
Warning

A124426-100G A124426-10G A124426-1G A124426-25G A124426-5G
77094-674EA 292.81 USD
77094-674 77094-676 77094-678 77094-680 77094-682
4-Chloro-2,3-dihydroinden-1-one 97%
4-Chloro-2,3-dihydroinden-1-one

Precaución: for Research Use Only

Formula: C₉H₇ClO
MW: 166.6000 g/mol
MDL Number: MFCD08544408
CAS Number: 15115-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR