3,4,5-Trichloropyridine 97%

Supplier: Ambeed
Warning

A124373-100G A124373-10G A124373-25G A124373-5G
77094-666EA 50.93 USD
77094-666 77094-668 77094-670 77094-672
3,4,5-Trichloropyridine 97%
3,4,5-Trichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₃N
MW: 182.44 g/mol
MDL Number: MFCD00051685
CAS Number: 33216-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR