RG7388 98%

Supplier: Ambeed
Warning

Synonyms: Idasanutlin, 4-((2R,3S,4R,5S)-3-(3-Chloro-2-fluorophenyl)-4-(4-chloro-2-fluorophenyl)-4-cyano-5-neopentylpyrrolidine-2-carboxamido)-3-methoxybenzoic acid

A124368-100MG A124368-1MG A124368-2MG A124368-5MG A124368-250MG A124368-1G A124368-50MG
77094-658EA 168.69 USD
77094-658 77094-660 77094-662 77094-664 77571-464 77611-062 77685-090
RG7388 98%
RG7388

Precaución: for Research Use Only

Formula: C₃₁H₂₉Cl₂F₂N₃O₄
MW: 616.48 g/mol
MDL Number: MFCD26142931
CAS Number: 1229705-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR