Fmoc-Met(O2)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Met(O₂)-OH, N-Fmoc-L-methionine sulphone

A123943-10G A123943-1G A123943-250MG A123943-5G A123943-25G
77094-634EA 49 USD
77094-634 77094-636 77094-638 77566-370 77685-087
Fmoc-Met(O2)-OH 95%
Fmoc-Met(O2)-OH

Precaución: For research use only.

Formula: C₂₀H₂₁NO₆S
MW: 403.449 g/mol
MDL Number: MFCD00153361
CAS Number: 163437-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR