Methyl 5-chloro-2-iodobenzoate 95%

Supplier: Ambeed
Warning

A123880-1G A123880-250MG A123880-25G A123880-5G
77094-626EA 15 USD
77094-626 77094-628 77094-630 77094-632
Methyl 5-chloro-2-iodobenzoate 95%
Methyl 5-chloro-2-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClIO₂
MDL Number: MFCD00144772
CAS Number: 289039-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR