4-Amino-3-chlorobenzoic acid 98%

Supplier: Ambeed
Warning

A123726-100G A123726-1G A123726-25G
77094-616EA 164.44 USD
77094-616 77094-618 77094-620
4-Amino-3-chlorobenzoic acid 98%
4-Amino-3-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD00460423
CAS Number: 2486-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR