(1-Tosyl-1H-indol-4-yl)methanamine 95%

Supplier: Ambeed
Warning

A123566-100MG A123566-250MG
77094-590EA 75.33 USD
77094-590 77094-592
(1-Tosyl-1H-indol-4-yl)methanamine 95%
(1-Tosyl-1H-indol-4-yl)methanamine

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₂S
MW: 300.38 g/mol
MDL Number: MFCD22573954
CAS Number: 1145678-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR