2-Bromo-4-chloro-6-nitrophenol 97%

Supplier: Ambeed
Danger

A123258-1G A123258-250MG A123258-25G A123258-5G
77094-574EA 21.99 USD
77094-574 77094-576 77094-578 77094-580
2-Bromo-4-chloro-6-nitrophenol 97%
2-Bromo-4-chloro-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₃
MDL Number: MFCD00024243
CAS Number: 15969-10-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR