2-Chloro-3-fluoro-5-methylpyridine 98%

Supplier: Ambeed
Warning

A123196-100G A123196-10G A123196-1G A123196-5G
77094-556EA 508.18 USD
77094-556 77094-558 77094-560 77094-562
2-Chloro-3-fluoro-5-methylpyridine 98%
2-Chloro-3-fluoro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD06658238
CAS Number: 34552-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR