2,4-Difluoro-5-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A123127-100MG A123127-1G A123127-250MG
77094-552EA 73 USD
77094-552 77901-532 77901-480
2,4-Difluoro-5-methoxyphenylboronic acid 98%
2,4-Difluoro-5-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.936 g/mol
MDL Number: MFCD21609510
CAS Number: 1395417-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR