D-Amygdalin 95%

Supplier: Ambeed
Warning

Synonyms: Laetrile, Amygdalin, Amygdaloside, D-Mandelonitrile 6-O-β-D-glucosido-β-D-glucoside, (R)-2-Phenyl-2-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)acetonitrile, D-Mandelonitrile-β-gentiobioside

A122268-10G A122268-1G A122268-250MG A122268-5G A122268-100MG A122268-25G
77094-532EA 15 USD
77094-532 77094-534 77094-536 77094-538 77685-077 77685-078
D-Amygdalin 95%
D-Amygdalin

Precaución: for Research Use Only

Formula: C₂₀H₂₇NO₁₁
MDL Number: MFCD00006598
CAS Number: 29883-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR