2-Amino-3-(pyridin-3-yl)propanoic acid 97%

Supplier: Ambeed
Danger

Synonyms: H-DL-3-Pal-OH, H-β-(3-Pyridyl)-DL-Ala-OH

A122246-10G A122246-1G A122246-250MG A122246-25G A122246-5G
77094-522EA 216.43 USD
77094-522 77094-524 77094-526 77094-528 77094-530
2-Amino-3-(pyridin-3-yl)propanoic acid 97%
2-Amino-3-(pyridin-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.1772 g/mol
MDL Number: MFCD02259525
CAS Number: 17470-24-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR