2-Chloro-3-nitropyridin-4-amine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-chloro-3-nitropyridine

A121905-100G A121905-10G A121905-1G A121905-25G A121905-5G
77094-512EA 233 USD
77094-512 77094-514 77094-516 77094-518 77094-520
2-Chloro-3-nitropyridin-4-amine 97%
2-Chloro-3-nitropyridin-4-amine

Precaución: for Research Use Only

Formula: C₅H₄ClN₃O₂
MW: 173.5572 g/mol
MDL Number: MFCD07368898
CAS Number: 2789-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR