5-Chloro-2-iodo-m-xylene 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2,6-dimethyliodobenzene

A121507-1G A121507-100MG
77094-494EA 116.7 USD
77094-494 77685-069
5-Chloro-2-iodo-m-xylene 97%
5-Chloro-2-iodo-m-xylene

Precaución: for Research Use Only

Formula: C₈H₈ClI
MW: 266.50 g/mol
MDL Number: MFCD11846954
CAS Number: 404337-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR