PD-1/PD-L1 Inhibitor 1 98%

Supplier: Ambeed
Warning

A121482-1MG A121482-25MG A121482-5MG A121482-50MG A121482-100MG
77094-488EA 16.98 USD
77094-488 77094-490 77094-492 77571-274 77611-032
PD-1/PD-L1 Inhibitor 1 98%
PD-1/PD-L1 Inhibitor 1

Precaución: for Research Use Only

Formula: C₂₉H₃₃NO₅
MW: 475.58 g/mol
MDL Number: MFCD28978741
CAS Number: 1675201-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR