2-Bromo-4-chloroaniline 97%

Supplier: Ambeed
Warning

A121355-100G A121355-10G A121355-25G A121355-500G A121355-5G
77094-468EA 50.93 USD
77094-468 77094-470 77094-472 77094-474 77094-476
2-Bromo-4-chloroaniline 97%
2-Bromo-4-chloroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrClN
MW: 206.47 g/mol
MDL Number: MFCD00041313
CAS Number: 873-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR