Calcium 4-(((2S,4R)-1-([1,1'-biphenyl]-4-yl)-5-ethoxy-4-methyl-5-oxopentan-2-yl)amino)-4-oxobutanoate 98%

Supplier: Ambeed
Danger

A120485-10MG A120485-50MG A120485-5G A120485-250MG A120485-25G
77094-428EA 5 USD
77094-428 77312-046 77312-048 77685-058 77685-059
Calcium 4-(((2S,4R)-1-([1,1'-biphenyl]-4-yl)-5-ethoxy-4-methyl-5-oxopentan-2-yl)amino)-4-oxobutanoate 98%
Calcium 4-(((2S,4R)-1-([1,1'-biphenyl]-4-yl)-5-ethoxy-4-methyl-5-oxopentan-2-yl)amino)-4-oxobutanoate

Precaución: for Research Use Only

Formula: C₄₈H₅₆CaN₂O₁₀
MDL Number: MFCD28137893
CAS Number: 1369773-39-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR