CL-387785 98%

Supplier: Ambeed
Warning

A119062-10MG A119062-25MG A119062-2MG A119062-50MG A119062-5MG A119062-100MG
77094-396EA 62 USD
77094-396 77094-398 77094-400 77094-402 77094-404 77094-406 77685-047
CL-387785 98%
CL-387785

Precaución: for Research Use Only

Formula: C₁₈H₁₃BrN₄O
MDL Number: MFCD02179194
CAS Number: 194423-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR