8-Methyl-4-quinolinol 95%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-8-methylquinoline, 8-Methylquinolin-4-ol

A118821-10G A118821-1G A118821-5G
77094-384EA 185.66 USD
77094-384 77094-386 77094-388
8-Methyl-4-quinolinol 95%
8-Methyl-4-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD00272344
CAS Number: 23432-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR