2-Bromo-6-nitrophenol 98%

Supplier: Ambeed
Warning

A118749-100G A118749-10G A118749-1G A118749-25G A118749-500G A118749-5G
77094-366EA 77.45 USD
77094-366 77094-368 77094-370 77094-372 77094-374 77094-376
2-Bromo-6-nitrophenol 98%
2-Bromo-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄BrNO₃
MDL Number: MFCD02683216
CAS Number: 13073-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR