7-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%

Supplier: Ambeed
Warning

A118577-100MG A118577-1G A118577-250MG A118577-5G
77094-356EA 70.02 USD
77094-356 77094-358 77094-360 77094-362
7-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
7-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO·ClH
MW: 199.677 g/mol
MDL Number: MFCD01444681
CAS Number: 1745-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR