1-(5-Bromo-2-hydroxy-4-methoxyphenyl)ethanone 97%

Supplier: Ambeed
Warning

A118343-1G A118343-250MG
77094-334EA 162.32 USD
77094-334 77094-336
1-(5-Bromo-2-hydroxy-4-methoxyphenyl)ethanone 97%
1-(5-Bromo-2-hydroxy-4-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
MDL Number: MFCD00597545
CAS Number: 39503-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR