3-(Propionyloxy)benzoic acid 97%

Supplier: Ambeed
Warning

A118221-1G A118221-250MG
77094-330EA 15 USD
77094-330 77094-332
3-(Propionyloxy)benzoic acid 97%
3-(Propionyloxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD20652409
CAS Number: 51988-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR