7-Bromochroman-4-ol 95%

Supplier: Ambeed
Warning

A117803-1G A117803-250MG A117803-5G
77094-294EA 83.82 USD
77094-294 77094-296 77094-298
7-Bromochroman-4-ol 95%
7-Bromochroman-4-ol

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD23806968
CAS Number: 18385-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR