(2-((2,5-Dichloropyrimidin-4-yl)amino)phenyl)dimethylphosphine oxide 96%

Supplier: Ambeed
Warning

A117681-10G A117681-250MG A117681-5G A117681-1G A117681-25G
77094-288EA 157 USD
77094-288 77094-290 77094-292 77571-064 77901-393
(2-((2,5-Dichloropyrimidin-4-yl)amino)phenyl)dimethylphosphine oxide 96%
(2-((2,5-Dichloropyrimidin-4-yl)amino)phenyl)dimethylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₂H₁₂Cl₂N₃OP
MDL Number: MFCD29477700
CAS Number: 1197953-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR