D-Serine benzyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Ser-OBzl·HCl, H-D-Ser-OBzl.HCl, Benzyl D-serinate HCl

A117391-10G A117391-25G A117391-1G A117391-100G A117391-5G
77094-276EA 47 USD
77094-276 77094-278 77610-964 77685-036 77685-037
D-Serine benzyl ester hydrochloride 97%
D-Serine benzyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃·HCl
MDL Number: MFCD00191017
CAS Number: 151651-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR