4-Amino-2,3-difluoro-5-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A117198-1G A117198-5G
77094-274EA 17.01 USD
77094-274 77571-030
4-Amino-2,3-difluoro-5-nitrobenzoic acid 95%
4-Amino-2,3-difluoro-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄F₂N₂O₄
MDL Number: MFCD08543937
CAS Number: 284030-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR