Ethyl 5,5,5-trifluoro-2,4-dioxopentanoate 95%

Supplier: Ambeed
Warning

A117089-100MG A117089-250MG A117089-5G
77094-268EA 36.08 USD
77094-268 77094-270 77094-272
Ethyl 5,5,5-trifluoro-2,4-dioxopentanoate 95%
Ethyl 5,5,5-trifluoro-2,4-dioxopentanoate

Precaución: for Research Use Only

Formula: C₇H₇F₃O₄
MW: 212.12 g/mol
MDL Number: MFCD08559044
CAS Number: 893643-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR