ZCL278 ≥98%

Supplier: Ambeed
Danger

Synonyms: 2-(4-Bromo-2-chlorophenoxy)-N-((4-(N-(4,6-dimethylpyrimidin-2-yl)sulphamoyl)phenyl)carbamothioyl)acetamide

A117081-10MG A117081-1MG A117081-25MG A117081-5MG
77094-260EA 26.53 USD
77094-260 77094-262 77094-264 77094-266
ZCL278 ≥98%
ZCL278

Precaución: for Research Use Only

Formula: C₂₁H₁₉BrClN₅O₄S₂
MW: 584.89 g/mol
MDL Number: MFCD03623090
CAS Number: 587841-73-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR