1-(2,6-Dichlorophenyl)oxindole 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2,6-Dichlorophenyl)-2-indolinone, 1-(2,6-Dichlorophenyl)-2-oxoindoline, Diclofenac Amide

A116302-100G A116302-10G A116302-25G A116302-500G A116302-5G
77094-236EA 23.34 USD
77094-236 77094-238 77094-240 77094-242 77094-244
1-(2,6-Dichlorophenyl)oxindole 97%
1-(2,6-Dichlorophenyl)oxindole

Precaución: for Research Use Only

Formula: C₁₄H₉Cl₂NO
MW: 278.13 g/mol
MDL Number: MFCD00451393
CAS Number: 15362-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR