2,2'-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)azanediyl)diacetic acid bis-4-methylbenzenesulfonic acid salt 97%

Supplier: Ambeed
Warning

A116217-100MG A116217-250MG
77094-232EA 140.04 USD
77094-232 77094-234
2,2'-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)azanediyl)diacetic acid bis-4-methylbenzenesulfonic acid salt 97%
2,2'-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)azanediyl)diacetic acid bis-4-methylbenzenesulfonic acid salt

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₂O₁₂S₂
MW: 600.62 g/mol
MDL Number: MFCD29905490
CAS Number: 2096496-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR