Methyl 2,5-Diaminobenzoate 95%

Supplier: Ambeed
Warning

A115493-100MG A115493-250MG
77094-210EA 64 USD
77094-210 77094-212
Methyl 2,5-Diaminobenzoate 95%
Methyl 2,5-Diaminobenzoate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD12913485
CAS Number: 49592-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR