Z-D-Gln(Trt)-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-5-Amino-2-(((benzyloxy)carbonyl)(trityl)amino)-5-oxopentanoic acid
A115397-10G
A115397-1G
77094-206EA
43
USD
77094-206
77094-208
Z-D-Gln(Trt)-OH 97%
Z-D-Gln(Trt)-OH
Precaución: for Research Use Only
Formula:
C₃₂H₃₀N₂O₅ |
MDL Number:
MFCD00237348 CAS Number: 200625-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...