(3S,11aR)-N-(2,4-Difluorobenzyl)-6-hydroxy-3-methyl-5,7-dioxo-2,3,5,7,11,11a-hexahydrooxazolo[3,2-a]pyrido[1,2-d]pyrazine-8-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: Cabotegravir

A115208-250MG A115208-100MG A115208-1G A115208-5G
77094-204EA 50.93 USD
77094-204 77549-272 77549-274 77570-892
(3S,11aR)-N-(2,4-Difluorobenzyl)-6-hydroxy-3-methyl-5,7-dioxo-2,3,5,7,11,11a-hexahydrooxazolo[3,2-a]pyrido[1,2-d]pyrazine-8-carboxamide 98%
(3S,11aR)-N-(2,4-Difluorobenzyl)-6-hydroxy-3-methyl-5,7-dioxo-2,3,5,7,11,11a-hexahydrooxazolo[3,2-a]pyrido[1,2-d]pyrazine-8-carboxamide

Precaución: for Research Use Only

Formula: C₁₉H₁₇F₂N₃O₅
MW: 405.35 g/mol
MDL Number: MFCD25976748
CAS Number: 1051375-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR