3-Iodo-7-methoxy-1H-pyrrolo[3,2-b]pyridine 95%

Supplier: Ambeed
Warning

A115089-100MG A115089-250MG
77094-200EA 77.45 USD
77094-200 77094-202
3-Iodo-7-methoxy-1H-pyrrolo[3,2-b]pyridine 95%
3-Iodo-7-methoxy-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₇IN₂O
MW: 274.06 g/mol
MDL Number: MFCD12963141
CAS Number: 1190318-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR