(3-Chloro-2-fluoro-5-methylphenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A114488-1G
A114488-250MG
A114488-5G
77094-178EA
47
USD
77094-178
77094-180
77094-182
(3-Chloro-2-fluoro-5-methylphenyl)boronic acid 98%
(3-Chloro-2-fluoro-5-methylphenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₇H₇BClFO₂ |
MDL Number:
MFCD22571810 CAS Number: 352535-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...