N-Benzyloxycarbonyl-L-alanine methyl ester 98%

Supplier: Ambeed
Warning

A114474-1G A114474-5G A114474-100G A114474-500G A114474-10G A114474-25G
77094-172EA 9 USD
77094-172 77094-174 77094-176 77685-014 77685-015 77900-508 77900-586
N-Benzyloxycarbonyl-L-alanine methyl ester 98%
N-Benzyloxycarbonyl-L-alanine methyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
MDL Number: MFCD00270464
CAS Number: 28819-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR