2-Chloro-6-nitroaniline 97%

Supplier: Ambeed
Warning

A114303-100G A114303-10G A114303-1G A114303-25G A114303-5G
77094-140EA 184.6 USD
77094-140 77094-142 77094-144 77094-146 77094-148
2-Chloro-6-nitroaniline 97%
2-Chloro-6-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
MDL Number: MFCD00179568
CAS Number: 769-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR