11-Aminoundecanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 11-Aminoundecylic acid

A113608-100G A113608-10G A113608-25G A113608-500G
77094-108EA 30.77 USD
77094-108 77094-110 77094-112 77094-114
11-Aminoundecanoic acid 97%
11-Aminoundecanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₃NO₂
MW: 201.31 g/mol
MDL Number: MFCD00008150
CAS Number: 2432-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR