1-Bromo-3,5-diisopropylbenzene 98%

Supplier: Ambeed
Warning

A112986-100MG A112986-250MG
77094-094EA 72.15 USD
77094-094 77094-096
1-Bromo-3,5-diisopropylbenzene 98%
1-Bromo-3,5-diisopropylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇Br
MW: 241.16 g/mol
MDL Number: MFCD12547791
CAS Number: 23058-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR