2-Bromo-4-chloro-5-nitrotoluene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-5-chloro-2-methyl-4-nitrobenzene

A112756-100MG A112756-250MG
77094-084EA 70 USD
77094-084 77094-086
2-Bromo-4-chloro-5-nitrotoluene 95%
2-Bromo-4-chloro-5-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MDL Number: MFCD00051512
CAS Number: 40371-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR