4-Bromo-5-chloro-2-methylaniline 97%

Supplier: Ambeed
Warning

A112520-1G A112520-25G A112520-5G
77094-056EA 6.37 USD
77094-056 77094-058 77094-060
4-Bromo-5-chloro-2-methylaniline 97%
4-Bromo-5-chloro-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MW: 220.5 g/mol
MDL Number: MFCD07782182
CAS Number: 30273-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR