3-Bromo-4-nitropyridine-N-oxide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-nitropyridine 1-oxide

A112290-5G A112290-25G
77094-044EA 43 USD
77094-044 77901-883
3-Bromo-4-nitropyridine-N-oxide 98%
3-Bromo-4-nitropyridine N-oxide

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₃
MW: 219.00 g/mol
MDL Number: MFCD00085925
CAS Number: 1678-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR