2-Bromo-6-chloro-4-fluoroaniline 97%

Supplier: Ambeed
Warning

A111978-100G A111978-10G A111978-1G A111978-250MG A111978-5G A111978-25G
77094-034EA 143 USD
77094-034 77094-036 77094-038 77094-040 77094-042 77684-993
2-Bromo-6-chloro-4-fluoroaniline 97%
2-Bromo-6-chloro-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MDL Number: MFCD00051707
CAS Number: 201849-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR