5-(2-(Methylthio)ethoxy)pyrimidin-2-amine 95%

Supplier: Ambeed
Warning

A111941-100MG A111941-250MG
77094-026EA 49.87 USD
77094-026 77094-028
5-(2-(Methylthio)ethoxy)pyrimidin-2-amine 95%
5-(2-(Methylthio)ethoxy)pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₇H₁₁N₃OS
MW: 185.25 g/mol
MDL Number: MFCD20694880
CAS Number: 1006599-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR