4-Nitro-1-indanone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Nitroindan-1-one

A111130-100MG A111130-1G A111130-250MG A111130-5G
77094-004EA 8 USD
77094-004 77094-006 77094-008 77901-929
4-Nitro-1-indanone 97%
4-Nitro-1-indanone

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MDL Number: MFCD06656901
CAS Number: 24623-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR