4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine 97%

Supplier: Ambeed
Warning

A110887-1G A110887-250MG A110887-5G
77093-996EA 13 USD
77093-996 77093-998 77901-030
4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine 97%
4-(Biphenyl-4-yl)-6-(4-bromophenyl)-2-phenylpyrimidine

Precaución: for Research Use Only

Formula: C₂₈H₁₉BrN₂
MDL Number: MFCD28975104
CAS Number: 1421599-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR