5-Bromo-4-methylthiophene-2-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Bromo-4-methylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A110509-100MG A110509-1G A110509-250MG A110509-5G
77093-984EA 37 USD
77093-984 77093-986 77093-988 77093-990
5-Bromo-4-methylthiophene-2-boronic acid, pinacol ester 98%
5-Bromo-4-methylthiophene-2-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₆BBrO₂S
MDL Number: MFCD12405479
CAS Number: 1111095-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR