3-Fluorostyrene 98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A110077-1G A110077-250MG A110077-25G A110077-5G
77093-962EA 22.28 USD
77093-962 77093-964 77093-966 77093-968
3-Fluorostyrene 98% stabilized
3-Fluorostyrene

Precaución: for Research Use Only

Formula: C₈H₇F
MW: 122.14 g/mol
MDL Number: MFCD00000341
CAS Number: 350-51-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR