4,6-Dichloro-2-(methylthio)pyrimidine-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A109784-1G A109784-250MG A109784-5G
77093-938EA 21.22 USD
77093-938 77093-940 77093-942
4,6-Dichloro-2-(methylthio)pyrimidine-5-carboxylic acid 98%
4,6-Dichloro-2-(methylthio)pyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂S
MW: 239.0800 g/mol
MDL Number: MFCD06208699
CAS Number: 313339-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR